Learn More
Sigma Aldrich Fine Chemicals Biosciences PKH26 Red fluorescent Cell
Catalog No. 502904287
Encompass
This kit provides enough dye for up to 50 samples of 20 million cells each and enough diluent for up to 30 samples of 20 million cells each when using a 2 mL final staining volume containing a final concentration of 2 x 10-6 M of PKH26 dye. Users must determine the optimal dye and cell concentrations from their cell type(s) and experimental purposes.
Catalog No. 50-290-4287 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MIDI261KT
This kit provides enough dye for up to 50 samples of 20 million cells each and enough diluent for up to 30 samples of 20 million cells each when using a 2 mL final staining volume containing a final concentration of 2 x 10-6 M of PKH26 dye. Users must determine the optimal dye and cell concentrations from their cell type(s) and experimental purposes.