Learn More
Enzo Life Sciences PKI (14-22) amide (myristoylated) (0.5 mg) CAS: 201422-03-9
SDP
Catalog No. NC1581641
Encompass_Preferred
PKA inhibitor. Heat-stable protein kinase inhibitor (PKI) peptide sequence (14-22). N-terminus is myristoylated to increase cell membrane permeability. The non-myristoylated version of this peptide is a highly specific inhibitor of cAMP-dependent protein kinase (PKA) with Ki=36 nM. This product is a useful tool for studying PKA in cellular systems. Alternative Name: cAMP dependent protein kinase inhibitor (14 22) amide (myristoylated). Formula: C53H100N20O12. MW: 1209.5. CAS: 201422-03-9. Formulation: Lyophilized. Trifluoroacetate salt. Purity: ≥95% (HPLC). Sequence N-myristoyl-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-NH2. Appearance: Solid. Solubility: Soluble to at least 1mg/ml in water. Long Term Storage: -20°C. Handling: Protect from moisture.
Catalog No. NC1581641 Supplier Enzo Life Sciences Supplier No. BMLP2100500
PKA inhibitor. Heat-stable protein kinase inhibitor (PKI) peptide sequence (14-22). N-terminus is myristoylated to increase cell membrane permeability. The non-myristoylated version of this peptide is a highly specific inhibitor of cAMP-dependent protein kinase (PKA) with Ki=36 nM. This product is a useful tool for studying PKA in cellular systems. Alternative Name: cAMP dependent protein kinase inhibitor (14 22) amide (myristoylated). Formula: C53H100N20O12. MW: 1209.5. CAS: 201422-03-9. Formulation: Lyophilized. Trifluoroacetate salt. Purity: ≥95% (HPLC). Sequence N-myristoyl-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-NH2. Appearance: Solid. Solubility: Soluble to at least 1mg/ml in water. Long Term Storage: -20°C. Handling: Protect from moisture.