Learn More
Medchemexpress LLC HY-P1291A 5mg , PKI 14-22 amide,myristoylated (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1291A5MG
Encompass_Preferred
HY-P1291A 5mg PKI 14-22 amide,myristoylated (TFA) CAS: PKI 14-22 amide,myristoylated TFA is a potent cAMP-dependent PKA inhibitor. PKI 14-22 amide,myristoylated TFA reduces the IgG-mediated phagocytic response and also inhibits neutrophil adhesion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0326