Learn More
Medchemexpress LLC Pki 14-22 amide,myristoylated | 201422-03-9 | 99.7% | 1209.49 | 1 MG
SDP
Catalog No. 5000424213
Encompass_Preferred
PKI 14-22 amide, myristoylated, is a selective, cAMP-dependent, competitive PKA inhibitor with a Ki of approximately 36 nM. Its myristoylation modification enhances its permeability to cell membranes and blood-brain barriers. This compound can inhibit the phosphorylation of cAMP-dependent downstream targets like CREB and is suitable for research in opioid tolerance mechanisms and antiviral drugs.
  • Selective, cAMP-dependent, competitive PKA inhibitor
  • Myristoylation enhances cell membrane and blood-brain barrier permeability
  • Inhibits phosphorylation of cAMP-dependent downstream targets (e.g., CREB)
  • Prevents morphine analgesic tolerance in mice
  • Inhibits Zika virus protein translation and RNA synthesis
Catalog No. 50-004-24213 Supplier Medchemexpress LLC Supplier No. HYP12911MG
PKI 14-22 amide, myristoylated, is a selective, cAMP-dependent, competitive PKA inhibitor with a Ki of approximately 36 nM. Its myristoylation modification enhances its permeability to cell membranes and blood-brain barriers. This compound can inhibit the phosphorylation of cAMP-dependent downstream targets like CREB and is suitable for research in opioid tolerance mechanisms and antiviral drugs.
  • Selective, cAMP-dependent, competitive PKA inhibitor
  • Myristoylation enhances cell membrane and blood-brain barrier permeability
  • Inhibits phosphorylation of cAMP-dependent downstream targets (e.g., CREB)
  • Prevents morphine analgesic tolerance in mice
  • Inhibits Zika virus protein translation and RNA synthesis