Learn More
Medchemexpress LLC PKI-166 10mg | 187724-61-4 | 330.38 | C20H18N4O | 10 MG
SDP
Catalog No. 5000308289
Encompass_Preferred
PKI-166 is a potent, selective, and orally bioavailable epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor with a reported IC50 of 0.7 nM. It is provided for research use only and has molecular formula C20H18N4O and high purity by HPLC.
  • Potent EGFR kinase inhibition (IC50 ≈ 0.7 nM).
  • High chemical purity (≥98% by HPLC).
  • Suitable for in vitro and in vivo research applications.
  • Orally bioavailable in preclinical studies.
  • Well-characterized molecular properties for experimental reproducibility.
Catalog No. 50-003-08289 Supplier Medchemexpress LLC Supplier No. HY11715510MG
PKI-166 is a potent, selective, and orally bioavailable epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor with a reported IC50 of 0.7 nM. It is provided for research use only and has molecular formula C20H18N4O and high purity by HPLC.
  • Potent EGFR kinase inhibition (IC50 ≈ 0.7 nM).
  • High chemical purity (≥98% by HPLC).
  • Suitable for in vitro and in vivo research applications.
  • Orally bioavailable in preclinical studies.
  • Well-characterized molecular properties for experimental reproducibility.