Learn More
TARGETMOL CHEMICALS INC PKI-166 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T165495MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PKI-166 is an oral inhibitor of EGF-R tyrosine kinase (IC500.7 nM) that is both effective and selective. PKI-166 can effectively inhibit the growth and metastasis of various human cancer cells including pancreatic cancer. purity: 99%
Catalog No. 50-257-7333