Learn More
Medchemexpress LLC PKI-179 hydrochloride | 1463510-35-1 | 99.66% | 525.00 | 10 MG
SDP
Catalog No. 5000343631
Encompass_Preferred
PKI-179 hydrochloride is a potent and orally active dual PI3K/mTOR inhibitor. It exhibits activity over various PI3K isoforms and mTOR, along with E545K and H1047R. This compound demonstrates anti-tumor activity in vivo.
  • Potent and orally active dual PI3K/mTOR inhibitor
  • Inhibits PI3K-α, PI3K-β, PI3K-γ, PI3K-δ and mTOR
  • Shows activity over E545K and H1047R
  • Exhibits anti-tumor activity in vivo
Catalog No. 50-003-43631 Supplier Medchemexpress LLC Supplier No. HY11080A10MM/1ML
PKI-179 hydrochloride is a potent and orally active dual PI3K/mTOR inhibitor. It exhibits activity over various PI3K isoforms and mTOR, along with E545K and H1047R. This compound demonstrates anti-tumor activity in vivo.
  • Potent and orally active dual PI3K/mTOR inhibitor
  • Inhibits PI3K-α, PI3K-β, PI3K-γ, PI3K-δ and mTOR
  • Shows activity over E545K and H1047R
  • Exhibits anti-tumor activity in vivo