Learn More
Medchemexpress LLC PKI-179 hydrochloride | 1463510-35-1 | 99.7% | 525.00 | 10 MG
SDP
Catalog No. 5000344256
Encompass_Preferred
PKI-179 hydrochloride is a potent and orally active dual PI3K/mTOR inhibitor. It exhibits inhibitory activity against PI3K-α, PI3K-β, PI3K-γ, PI3K-δ, and mTOR, with varying IC50 values. This compound also shows activity against E545K and H1047R mutations and demonstrates anti-tumor effects in vivo.
  • Potent and orally active.
  • Dual PI3K/mTOR inhibitor.
  • Inhibits PI3K-α, PI3K-β, PI3K-γ, PI3K-δ, and mTOR.
  • Active against E545K and H1047R.
  • Shows anti-tumor activity in vivo.
Catalog No. 50-003-44256 Supplier Medchemexpress LLC Supplier No. HY11080A100MG
PKI-179 hydrochloride is a potent and orally active dual PI3K/mTOR inhibitor. It exhibits inhibitory activity against PI3K-α, PI3K-β, PI3K-γ, PI3K-δ, and mTOR, with varying IC50 values. This compound also shows activity against E545K and H1047R mutations and demonstrates anti-tumor effects in vivo.
  • Potent and orally active.
  • Dual PI3K/mTOR inhibitor.
  • Inhibits PI3K-α, PI3K-β, PI3K-γ, PI3K-δ, and mTOR.
  • Active against E545K and H1047R.
  • Shows anti-tumor activity in vivo.