Learn More
Cayman Chemical PKI-179 (hydrochloride), Purity: ≥98%, CAS: 1463510-35-1, 10 mg
SDP
Catalog No. 502981026
Encompass_Preferred
A dual PI3K and mTOR inhibitor; inhibits PI3K (IC50s = 8, 24, 17, and 74 nM for isoforms α, β, δ, and γ, respectively); inhibits PI3Kα mutants, E545K and H1047R (IC50s = 14 and 11 nM, respectively); inhibits mTOR (IC50 = 0.42 nM); antiproliferative against MDA-361 (breast) and PC3MM2 (prostate) cell lines (IC50s = 22 and 29 nM, respectively); inhibits tumor growth in MDA-361 mouse xenografts (50 mg/kg)
Catalog No. 50-298-1026 Supplier Cayman Chemical Supplier No. 2120210 MG
A dual PI3K and mTOR inhibitor; inhibits PI3K (IC50s = 8, 24, 17, and 74 nM for isoforms α, β, δ, and γ, respectively); inhibits PI3Kα mutants, E545K and H1047R (IC50s = 14 and 11 nM, respectively); inhibits mTOR (IC50 = 0.42 nM); antiproliferative against MDA-361 (breast) and PC3MM2 (prostate) cell lines (IC50s = 22 and 29 nM, respectively); inhibits tumor growth in MDA-361 mouse xenografts (50 mg/kg)