Learn More
Selleck Chemical LLC PKI-402 S2739-5mg
SDP
Catalog No. 503112599
Encompass_Preferred
PKI-402 is a potent dual pan-PI3K/mTOR inhibitor targeting PI3K / / / and mTOR with IC50 of 2 nM/7 nM/16 nM/14 nM and 3 nM respectively also potent to PI3K mutants E545K and H1047R
Catalog No. 50-311-2599 Supplier Selleck Chemical LLC Supplier No. S27395MG
PKI-402 is a potent dual pan-PI3K/mTOR inhibitor targeting PI3K / / / and mTOR with IC50 of 2 nM/7 nM/16 nM/14 nM and 3 nM respectively also potent to PI3K mutants E545K and H1047R