Learn More
Medchemexpress LLC HY-10683 5mg Medchemexpress, PKI-402 CAS:1173204-81-3 Purity:>98%
SDP
Catalog No. 501968355
Encompass_Preferred
Medchemexpress, HY-10683 5mg PKI-402 CAS:1173204-81-3 PKI-402 is a selective, reversible, ATP-competitive inhibitor of PI3K, including PI3K-α mutants, and mTOR (IC50=2, 3, 7,14 and 16 nM for PI3Kα, mTOR, PI3Kβ, PI3Kδ and PI3Kγ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8355 Supplier Medchemexpress LLC Supplier No. HY106835MG
Medchemexpress, HY-10683 5mg PKI-402 CAS:1173204-81-3 PKI-402 is a selective, reversible, ATP-competitive inhibitor of PI3K, including PI3K-α mutants, and mTOR (IC50=2, 3, 7,14 and 16 nM for PI3Kα, mTOR, PI3Kβ, PI3Kδ and PI3Kγ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.