Learn More
TARGETMOL CHEMICALS INC PKI-402 5MG
SDP
Catalog No. 502576894
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. PKI-402 is a potent PI3K and mTOR inhibitor. PKI-402 inhibits PI3K(alpha) mTOR PI3K(beta) PI3K(delta) and PI3K(gamma) with IC50s of 2 3 7 14 and 16 nM respectively. purity: 100%
Catalog No. 50-257-6894 Supplier TARGETMOL CHEMICALS INC Supplier No. T36565MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. PKI-402 is a potent PI3K and mTOR inhibitor. PKI-402 inhibits PI3K(alpha) mTOR PI3K(beta) PI3K(delta) and PI3K(gamma) with IC50s of 2 3 7 14 and 16 nM respectively. purity: 100%