Learn More
Enzo Life Sciences PKI (5-24) (0.5mg). CAS: 99534-03-9
SDP
Supplier:  Enzo Life Sciences BMLP2030500
Encompass_Preferred
Sequence is from the heat-stable skeletal muscle inhibitor protein of PKA. Peptide binds to the catalytic subunit of PKA and displaces the regulatory subunit. Mimics protein substrate by binding to the catalytic site via the Arg-cluster basic residues. Ki =2.3 nM for PKA. Alternative name: cAMP dependent protein kinase inhibitor (5 24). Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1280