Learn More
Medchemexpress LLC PKI(5-24) | 99534-03-9 | 99.99% | 2222.38 | 10 MG
SDP
Catalog No. 5000425733
Encompass_Preferred
PKI(5-24) is a potent, competitive, and synthetic peptide inhibitor of PKA (cAMP-dependent protein kinase), with a Ki of 2.3 nM. It corresponds to residues 5-24 in the naturally occurring heat-stable protein kinase inhibitor. This 20-residue peptide inhibits phosphotransferase activity of the mutant cGMP kinase with higher potency (Ki of 42 μM) than that of the wild type (Ki of 160 μM). This product is for research use only.
  • Potent, competitive, and synthetic peptide inhibitor of PKA (cAMP-dependent protein kinase)
  • Ki of 2.3 nM
  • Corresponds to residues 5-24 in the naturally occurring heat-stable protein kinase inhibitor
  • 20-residue peptide synthesized to correspond to the active site of the skeletal muscle inhibitor protein
  • Inhibits phosphotransferase activity of the mutant cGMP kinase with higher potency
  • For research use only
Catalog No. 50-004-25733 Supplier Medchemexpress LLC Supplier No. HYP022210MG
PKI(5-24) is a potent, competitive, and synthetic peptide inhibitor of PKA (cAMP-dependent protein kinase), with a Ki of 2.3 nM. It corresponds to residues 5-24 in the naturally occurring heat-stable protein kinase inhibitor. This 20-residue peptide inhibits phosphotransferase activity of the mutant cGMP kinase with higher potency (Ki of 42 μM) than that of the wild type (Ki of 160 μM). This product is for research use only.
  • Potent, competitive, and synthetic peptide inhibitor of PKA (cAMP-dependent protein kinase)
  • Ki of 2.3 nM
  • Corresponds to residues 5-24 in the naturally occurring heat-stable protein kinase inhibitor
  • 20-residue peptide synthesized to correspond to the active site of the skeletal muscle inhibitor protein
  • Inhibits phosphotransferase activity of the mutant cGMP kinase with higher potency
  • For research use only