Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pkiota-in-2 formic | 99.04% | 402.36 | 10 MG
SDP

Catalog No. 5000438700
Encompass_Preferred

PKCiota-IN-2 formic is a potent inhibitor of PKCiota (IC50 of 2.8 nM), and also inhibits PKC-α (IC50 of 71 nM) and PKC-ε (IC50 of 350 nM). This compound is for research use only.

  • Molecular weight of 402.36
  • Formula: C25H23N5O3
  • Appearance as a solid
  • Color ranges from light yellow to yellow
  • Soluble in DMSO at 100 mg/mL (248.53 mM); requires ultrasonic
  • Ships at room temperature in continental US
  • Store at 4°C sealed, away from moisture and light. In solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-38700 Supplier Medchemexpress LLC Supplier No. HY122858A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PKCiota-IN-2 formic is a potent inhibitor of PKCiota (IC50 of 2.8 nM), and also inhibits PKC-α (IC50 of 71 nM) and PKC-ε (IC50 of 350 nM). This compound is for research use only.

  • Molecular weight of 402.36
  • Formula: C25H23N5O3
  • Appearance as a solid
  • Color ranges from light yellow to yellow
  • Soluble in DMSO at 100 mg/mL (248.53 mM); requires ultrasonic
  • Ships at room temperature in continental US
  • Store at 4°C sealed, away from moisture and light. In solvent, store at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.