Learn More
Aviva Systems Biology Corp PKLR Antibody - N-terminal region (ARP41699_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41699_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PKLR. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PKLR. Alternate names: PK1, PKL, RPK, PKR, PKRLApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57959