Learn More
Biomatik Corporation Pkm Monoclonal Antibody
Supplier:  Biomatik Corporation CAC10273
Encompass_Preferred
Pkm Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to PKM. The Pkm Monoclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF, FC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Recombinant Human Pyruvate Kinase Pkm Protein (2-531Aa). Applications: ELISA, WB, IHC, IF, FC, IP. Target Synonyms: PKM. Accession Number: P14618. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7752