Learn More
Aviva Systems Biology Corp PKM2 Antibody - N-terminal region (ARP57788_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57788_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PKM2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PKM2. Alternate names: CTHBP, MGC3932, OIP3, PK3, PKM, TCB, THBP1, PKM2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-377