Learn More
Aviva Systems Biology Corp PKM2 Antibody - middle region (ARP57789_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57789_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PKM2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PKM2. Alternate names: CTHBP, MGC3932, OIP3, PK3, PKM, TCB, THBP1, PKM2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-286-378