Learn More
Medchemexpress LLC HY-103617 5mg Medchemexpress, PKM2-IN-1 CAS:94164-88-2 Purity:>98%
SDP
Catalog No. 501968143
Encompass_Preferred
Medchemexpress, HY-103617 5mg PKM2-IN-1 CAS:94164-88-2 PKM2-IN-1 is a pyruvate kinase M2 (PKM2) inhibitor with an IC50 of 2.95 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8143 Supplier Medchemexpress LLC Supplier No. HY1036175MG
Medchemexpress, HY-103617 5mg PKM2-IN-1 CAS:94164-88-2 PKM2-IN-1 is a pyruvate kinase M2 (PKM2) inhibitor with an IC50 of 2.95 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.