Learn More
Selleck Chemical LLC PKM2 inhibitor
SDP
Supplier:  Selleck Chemical LLC S861625MG
Encompass_Preferred
PKM2 inhibitor(Compound 3K PKM2-IN-1) displays PKM2 inhibitory activity with the IC50 value of 2 95 M The IC50 value for PKM1 is 4-5-fold higher than that for PKM2
Catalog No. 50-000-01739