Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PKN2 (PRK2), GST Tag Recombinant Protein

Catalog No. PV3879
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3879 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV3879 Supplier Invitrogen™ Supplier No. PV3879
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.

PKN2 (PRK2), a close relative of the PKCs, is a regulator of mitosis and exit from cytokinesis. Plays a role in the regulation of cell cycle progression, actin cytoskeleton assembly, cell migration, cell adhesion, tumor cell invasion and transcription activation signaling processes. Phosphorylates CTTN in hyaluronan-induced astrocytes and hence decreases CTTN ability to associate with filamentous actin. Phosphorylates HDAC5, therefore lead to impair HDAC5 import. Direct RhoA target required for the regulation of the maturation of primordial junctions into apical junction formation in bronchial epithelial cells. Required for G2/M phases of the cell cycle progression and abscission during cytokinesis in a ECT2-dependent manner. Stimulates FYN kinase activity that is required for establishment of skin cell-cell adhesion during keratinocytes differentiation. Regulates epithelial bladder cells speed and direction of movement during cell migration and tumor cell invasion. Inhibits Akt pro-survival-induced kinase activity. Mediates Rho protein-induced transcriptional activation via the c-fos serum response factor (SRF). Phosphorylates HCV NS5B leading to stimulation of HCV RNA replication.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16513
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.5mM EDTA, 2mM DTT, 150mM NaCl, 50% glycerol, 0.02% Triton X-100 and no preservative; pH 7.5
Gene ID (Entrez) 5586
Molecular Weight (g/mol) 141.5 kDa
Name Human PKN2 (PRK2), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 6030436C20Rik; AI507382; cardiolipin-activated protein kinase Pak2; p140 kinase; PAK2; Pak-2; PKN gamma; PKN2; PRK2; PRKCL2; PRO2042; protease-activated kinase 2; protein kinase C-like 2; protein kinase N2; protein-kinase C-related kinase 2; RP5-905H16.1; serine/threonine kinase 7; serine/threonine-protein kinase N2; serine/threonine-protein kinase N2; serine/threonine-protein kinase N2-like; si:ch211-83f14.1; Stk7; wu:fa97c07
Common Name PKN2
Gene Symbol PKN2
Kinase Family PKN Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage Storage Buffer: 50 mM Tris, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Mass 10 μg
Kinase Group Ser⁄Thr Kinases: ACG Group
Shipping Condition Dry Ice
Tag Position N-Terminal
Target Gene protein kinase N2
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Drug Discovery: Signal Transduction
Purity or Quality Grade 80% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.