Learn More
Cayman Chemical PKN3 InhIbItr Compound 16 25mg
SDP
Catalog No. 502942671
Encompass_Preferred
A serine/threonine protein kinase N3 inhibitor (IC50 = 14 nM); selective for this kinase over a panel of 234 kinases but does inhibit NLK, ACTR1, and RIPK2 at 1 µM, as well as GAK (IC50 = 74 nM in a BRET assay); inhibits viral replication in dengue virus-infected Huh7 cells (EC50 = 3.6 µM) without inducing cell death (CC50 = >10 µM)
Catalog No. 50-294-2671 Supplier Cayman Chemical Supplier No. 4143725 MG
A serine/threonine protein kinase N3 inhibitor (IC50 = 14 nM); selective for this kinase over a panel of 234 kinases but does inhibit NLK, ACTR1, and RIPK2 at 1 µM, as well as GAK (IC50 = 74 nM in a BRET assay); inhibits viral replication in dengue virus-infected Huh7 cells (EC50 = 3.6 µM) without inducing cell death (CC50 = >10 µM)