Learn More
ABclonal Technology PKNOX1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A153011000UL
Encompass_Preferred
Enables sequence-specific double-stranded DNA binding activity. Predicted to be involved in angiogenesis and regulation of transcription by RNA polymerase II. Predicted to act upstream of or within camera-type eye development, hemopoiesis, and positive regulation of transcription by RNA polymerase II. Predicted to be located in cytoplasm and nucleus. Predicted to be part of chromatin.
Catalog No. 50-001-05047