Learn More
Aviva Systems Biology Corp PKNOX2 Antibody - N-terminal region (ARP33366_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33366_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PKNOX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PKNOX2. Alternate names: PREP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-54983