Learn More
Aviva Systems Biology Corp PKP4 Antibody - N-terminal region (ARP64810_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64810_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PKP4. It was validated on Western Blot. . Alternate names: FLJ31261, FLJ42243, p0071ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-374