Learn More
Eurofins Discoverx PKR, active, 250 µg. (14-955M)
Supplier:  Eurofins Discoverx 14955M
Encompass
N-terminal GST-tagged and C-terminal 10His tagged recombinant, human PKR amino acids 252-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9479