Learn More
Carnabio Usa Inc PKR(EIF2AK2)/10UG/CATALYTIC
Supplier:  Carnabio Usa Inc 0515610
Encompass
PKR(EIF2AK2), Catalytic domain, Wild type, Amino Acid 252-551, NP_002750.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1063