Learn More
Medchemexpress LLC HY-13977A 10mg Medchemexpress, PKR-IN-C16 CAS:608512-97-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13977A10MG
Encompass_Preferred
Medchemexpress, HY-13977A 10mg PKR-IN-C16 CAS:608512-97-6 PKR-IN-C16 is a specific protein kinase (PKR) inhibitor. PKR-IN-C16 is able to inhibit the autophosphorylation of PKR and unlock the translation blockade induced by PKR in primary neuronal cultures. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9892