Learn More
Medchemexpress LLC PKR-IN-C51 | 1314594-23-4 | 99.8% | 367.45 | C23H21N5 | 50 MG
SDP
Catalog No. 5000228194
Encompass_Preferred
PKR-IN-C51 is an ATP-competitive inhibitor of double-stranded RNA-activated protein kinase (PKR) supplied for laboratory research use. It inhibits intracellular PKR activation in a dose-dependent manner and is provided as a solid powder with solution formats available.
  • Inhibits PKR activation in cells in a dose-dependent manner.
  • Acts via an ATP-competitive mechanism.
  • Available as a solid powder and as 10 mM solutions in DMSO.
  • Supplied in multiple pack sizes, including 50 mg.
  • Includes documentation: data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-28194 Supplier Medchemexpress LLC Supplier No. HY11813150MG
PKR-IN-C51 is an ATP-competitive inhibitor of double-stranded RNA-activated protein kinase (PKR) supplied for laboratory research use. It inhibits intracellular PKR activation in a dose-dependent manner and is provided as a solid powder with solution formats available.
  • Inhibits PKR activation in cells in a dose-dependent manner.
  • Acts via an ATP-competitive mechanism.
  • Available as a solid powder and as 10 mM solutions in DMSO.
  • Supplied in multiple pack sizes, including 50 mg.
  • Includes documentation: data sheet, certificate of analysis, and safety data sheet.