Learn More
Medchemexpress LLC Pkr-in-c51 | 1314594-23-4 | 99.8% | 367.45 g·mol⁻¹ | C23H21N5 | 10 MG
SDP
Catalog No. 5000226569
Encompass_Preferred
PKR-IN-C51 is an ATP-competitive inhibitor of double-stranded RNA-activated protein kinase (PKR) with a reported IC50 of approximately 9 μM. It is supplied as a high-purity solid intended for research use and is stored protected from light.
  • High purity (99.8%).
  • ATP-competitive PKR inhibition, IC50 ≈ 9 μM.
  • Molecular formula C23H21N5; molecular weight 367.45 g·mol⁻¹.
  • Supplied as a 10 mg solid for laboratory research use.
  • Store protected from light; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-26569 Supplier Medchemexpress LLC Supplier No. HY11813110MG
PKR-IN-C51 is an ATP-competitive inhibitor of double-stranded RNA-activated protein kinase (PKR) with a reported IC50 of approximately 9 μM. It is supplied as a high-purity solid intended for research use and is stored protected from light.
  • High purity (99.8%).
  • ATP-competitive PKR inhibition, IC50 ≈ 9 μM.
  • Molecular formula C23H21N5; molecular weight 367.45 g·mol⁻¹.
  • Supplied as a 10 mg solid for laboratory research use.
  • Store protected from light; in solvent: -80°C (6 months), -20°C (1 month).