Learn More
Medchemexpress LLC Pkr-in-c51 | 1314594-23-4 | 99.8% | 367.45 g/mol | C23H21N5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1181315MG
Encompass_Preferred
PKR-IN-C51 is an ATP-competitive inhibitor of double-stranded RNA-activated protein kinase (PKR) for research use in cellular and biochemical studies. It has a reported IC50 of 9 μM against PKR, a molecular weight of 367.45 g/mol, and is supplied at high purity for laboratory applications.
  • ATP-competitive inhibitor of PKR for cellular and biochemical studies.
  • Reported IC50 of 9 μM against PKR.
  • High supplied purity (≈99.8%).
  • Available in small research quantities, including 5 mg.
  • Storage: in solvent, -80°C for 6 months or -20°C for 1 month; protect from light.
Catalog No. 50-002-28814