Learn More
Apexbio Technology LLC PKR Inhibitor 608512-97-6 5mg
SDP
Catalog No. 502240172
Encompass_Preferred
PKR Inhibitor (CAS 608512-97-6) is a small molecule that selectively inhibits double-stranded RNA-dependent protein kinase (PKR) a critical regulator of translational initiation via kinase-dependent phosphorylation events With reported IC50 values of 186 210 nM this inhibitor blocks PKR autophosphorylation thereby alleviating PKR-mediated translational repression in vitro In SH-SY5Y cells PKR Inhibitor mitigates Ab-induced caspase 3 activation and protects against endoplasmic reticulum stress-associated neuronal cell death In animal models systemic administration suppresses the apoptotic PKR/eIF2 pathway without activating the mTOR/p70S6K cascade PKR Inhibitor is utilized in studies of translational control stress responses and neuroprotection mechanisms
This item is not returnable. View return policy
Catalog No. 50-224-0172 Supplier Apexbio Technology LLC Supplier No. C44395
PKR Inhibitor (CAS 608512-97-6) is a small molecule that selectively inhibits double-stranded RNA-dependent protein kinase (PKR) a critical regulator of translational initiation via kinase-dependent phosphorylation events With reported IC50 values of 186 210 nM this inhibitor blocks PKR autophosphorylation thereby alleviating PKR-mediated translational repression in vitro In SH-SY5Y cells PKR Inhibitor mitigates Ab-induced caspase 3 activation and protects against endoplasmic reticulum stress-associated neuronal cell death In animal models systemic administration suppresses the apoptotic PKR/eIF2 pathway without activating the mTOR/p70S6K cascade PKR Inhibitor is utilized in studies of translational control stress responses and neuroprotection mechanisms