Learn More
Cayman Chemical PKR Inhibitor, Purity: ≥95%, CAS: 608512-97-6, 1 mg
SDP
Catalog No. 502972968
Encompass_Preferred
Binds the ATP-binding site of PKR and blocks autophosphorylation (IC50 = 186-210 nM); protects neuroblastoma cells against cell damage triggered by ER stress; prevents phosphorylation of FADD, preventing apoptosis; reduces phosphorylation of PKR and eIF2α in the brain, enhancing long-term memory storage
Catalog No. 50-297-2968 Supplier Cayman Chemical Supplier No. 153231 MG
Binds the ATP-binding site of PKR and blocks autophosphorylation (IC50 = 186-210 nM); protects neuroblastoma cells against cell damage triggered by ER stress; prevents phosphorylation of FADD, preventing apoptosis; reduces phosphorylation of PKR and eIF2α in the brain, enhancing long-term memory storage