Learn More
Cayman Chemical PKR InhIbItr 5mg
SDP
Catalog No. 502973264
Encompass_Preferred
Binds the ATP-binding site of PKR and blocks autophosphorylation (IC50 = 186-210 nM); protects neuroblastoma cells against cell damage triggered by ER stress; prevents phosphorylation of FADD, preventing apoptosis; reduces phosphorylation of PKR and eIF2α in the brain, enhancing long-term memory storage
Catalog No. 50-297-3264 Supplier Cayman Chemical Supplier No. 153235 MG
Binds the ATP-binding site of PKR and blocks autophosphorylation (IC50 = 186-210 nM); protects neuroblastoma cells against cell damage triggered by ER stress; prevents phosphorylation of FADD, preventing apoptosis; reduces phosphorylation of PKR and eIF2α in the brain, enhancing long-term memory storage