Learn More
Medchemexpress LLC Pksi-527 | 128837-71-8 | ≥95.0% | 474.0 g/mol | C25H31N3O4 · HCl | 100MG
SDP
Catalog No. 5000237940
Encompass_Preferred
PKSI-527 is a small-molecule inhibitor of plasma kallikrein used in preclinical research to study the kallikrein-kinin system and inflammatory pathways. It exhibits a reported Ki of 0.81 μM and has been shown to suppress collagen-induced arthritis in animal models. The compound is typically supplied as the monohydrochloride salt (CAS 128837-71-8) and is intended for biochemical and pharmacology studies.
Catalog No. 50-002-37940 Supplier Medchemexpress LLC Supplier No. HY123133100MG
PKSI-527 is a small-molecule inhibitor of plasma kallikrein used in preclinical research to study the kallikrein-kinin system and inflammatory pathways. It exhibits a reported Ki of 0.81 μM and has been shown to suppress collagen-induced arthritis in animal models. The compound is typically supplied as the monohydrochloride salt (CAS 128837-71-8) and is intended for biochemical and pharmacology studies.