Learn More
Medchemexpress LLC Pksi-527 | 128837-71-8 | MFCD01745772 | 98.1% | 473.99 g·mol-1 | C25H32ClN3O4 | 10MG
SDP
Catalog No. 5000237373
Encompass_Preferred
PKSI-527 is a selective small-molecule inhibitor of plasma kallikrein used as a research reagent. It has been reported to suppress collagen-induced arthritis by modulating the kallikrein-kinin system and is commonly used in studies of coagulation, inflammation, and immunology. The compound is supplied as a solid (powder) with typical solubility in DMSO and is intended for laboratory research use only.
  • Selective plasma kallikrein inhibition for target validation studies.
  • Demonstrated activity in collagen-induced arthritis models.
  • High purity suitable for in vitro assays (purity approximately 98.1%).
  • Available in multiple package sizes to support different experiment scales.
  • Soluble in DMSO for assay formulation and screening.
Catalog No. 50-002-37373 Supplier Medchemexpress LLC Supplier No. HY12313310MG
PKSI-527 is a selective small-molecule inhibitor of plasma kallikrein used as a research reagent. It has been reported to suppress collagen-induced arthritis by modulating the kallikrein-kinin system and is commonly used in studies of coagulation, inflammation, and immunology. The compound is supplied as a solid (powder) with typical solubility in DMSO and is intended for laboratory research use only.
  • Selective plasma kallikrein inhibition for target validation studies.
  • Demonstrated activity in collagen-induced arthritis models.
  • High purity suitable for in vitro assays (purity approximately 98.1%).
  • Available in multiple package sizes to support different experiment scales.
  • Soluble in DMSO for assay formulation and screening.