Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PKSI-527 | 128837-71-8 | MFCD01745772 | >98.0% | 473.99 g/mol | C25H32ClN3O4 | 10MM 1ML
SDP

Catalog No. 5000237374
Encompass_Preferred

PKSI-527 is a selective small-molecule inhibitor of plasma kallikrein used in biochemical and pharmacological research. It has been reported to suppress collagen-induced arthritis by modulating the kallikrein-kinin system and is supplied as a hydrochloride salt suitable for in vitro and in vivo studies.

  • Selective plasma kallikrein inhibitor.
  • Demonstrated activity in collagen-induced arthritis models.
  • Supplied as the hydrochloride salt for improved stability and handling.
  • High solubility in DMSO; typical stock solutions prepared at 1 mM, 5 mM, or 10 mM.
  • Storage recommendations for powder and solutions to preserve stability.
  • Reported purity greater than 98% by some suppliers.
  • Molecular weight 473.99 g/mol; chemical formula C25H32ClN3O4.

Catalog No. 50-002-37374 Supplier Medchemexpress LLC Supplier No. HY12313310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PKSI-527 is a selective small-molecule inhibitor of plasma kallikrein used in biochemical and pharmacological research. It has been reported to suppress collagen-induced arthritis by modulating the kallikrein-kinin system and is supplied as a hydrochloride salt suitable for in vitro and in vivo studies.

  • Selective plasma kallikrein inhibitor.
  • Demonstrated activity in collagen-induced arthritis models.
  • Supplied as the hydrochloride salt for improved stability and handling.
  • High solubility in DMSO; typical stock solutions prepared at 1 mM, 5 mM, or 10 mM.
  • Storage recommendations for powder and solutions to preserve stability.
  • Reported purity greater than 98% by some suppliers.
  • Molecular weight 473.99 g/mol; chemical formula C25H32ClN3O4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.