Learn More
Medchemexpress LLC PKSI-527 hydrochloride | 128837-71-8 | MFCD01745772 | 98.1% | 473.99 g·mol⁻¹ | C25H32ClN3O4 | 50MG
SDP
Catalog No. 5000235753
Encompass_Preferred
PKSI-527 is a selective small-molecule plasma kallikrein inhibitor used in preclinical research to investigate the kallikrein-kinin pathway and inflammatory disease models such as collagen-induced arthritis. It is supplied as a characterized hydrochloride salt at high purity for laboratory use only and is intended for in vitro and in vivo research under appropriate approvals.
  • Highly selective plasma kallikrein inhibition.
  • Applicable to preclinical models of inflammatory disease.
  • Suitable for in vitro enzyme and cellular assays.
  • Appropriate for in vivo disease-model investigations.
  • Supplied at high purity for reproducible results.
  • Characterized by molecular formula and molecular weight.
  • Provided as a hydrochloride salt for stability and handling.
  • Intended for research use only; not for human consumption.
Catalog No. 50-002-35753 Supplier Medchemexpress LLC Supplier No. HY12313350MG
PKSI-527 is a selective small-molecule plasma kallikrein inhibitor used in preclinical research to investigate the kallikrein-kinin pathway and inflammatory disease models such as collagen-induced arthritis. It is supplied as a characterized hydrochloride salt at high purity for laboratory use only and is intended for in vitro and in vivo research under appropriate approvals.
  • Highly selective plasma kallikrein inhibition.
  • Applicable to preclinical models of inflammatory disease.
  • Suitable for in vitro enzyme and cellular assays.
  • Appropriate for in vivo disease-model investigations.
  • Supplied at high purity for reproducible results.
  • Characterized by molecular formula and molecular weight.
  • Provided as a hydrochloride salt for stability and handling.
  • Intended for research use only; not for human consumption.