Learn More
Medchemexpress LLC PKUMDL-LC-101-D04 100mg | 2143896-83-5 | 378.9 | C14H22N4O2S2 · HCl | 100 MG
SDP
Catalog No. 5000226641
Encompass_Preferred
PKUMDL-LC-101-D04 (GPX4-Activator-1d4) is a small-molecule allosteric activator of glutathione peroxidase 4 (GPX4) used in research to modulate ferroptosis and lipid-peroxide-mediated cell death. It increases GPX4 enzymatic activity in biochemical assays and has demonstrated activity in cell extracts and cellular protection assays.
  • Allosteric activator of glutathione peroxidase 4.
  • Increases GPX4 activity to about 150% in a cell-free assay at 20 μM.
  • Restores GPX4 activity in cell extracts at 61 μM.
  • Reduces cholesterol hydroperoxide-induced cell death in mouse embryonic fibroblasts.
  • High purity research-grade material suitable for biochemical and cellular assays.
  • Molecular weight 378.9 and formula C14H22N4O2S2 · HCl.
Catalog No. 50-002-26641 Supplier Medchemexpress LLC Supplier No. HY115627100MG
PKUMDL-LC-101-D04 (GPX4-Activator-1d4) is a small-molecule allosteric activator of glutathione peroxidase 4 (GPX4) used in research to modulate ferroptosis and lipid-peroxide-mediated cell death. It increases GPX4 enzymatic activity in biochemical assays and has demonstrated activity in cell extracts and cellular protection assays.
  • Allosteric activator of glutathione peroxidase 4.
  • Increases GPX4 activity to about 150% in a cell-free assay at 20 μM.
  • Restores GPX4 activity in cell extracts at 61 μM.
  • Reduces cholesterol hydroperoxide-induced cell death in mouse embryonic fibroblasts.
  • High purity research-grade material suitable for biochemical and cellular assays.
  • Molecular weight 378.9 and formula C14H22N4O2S2 · HCl.