Learn More
Medchemexpress LLC PKUMDL-LC-101-D04 10mM 1mL | 2143896-83-5 | 1 ML
SDP
Catalog No. 5000307946
Encompass_Preferred
PKUMDL-LC-101-D04 is a small-molecule allosteric activator of glutathione peroxidase 4 (GPX4) used in research to inhibit ferroptosis and modulate inflammatory responses. The compound is provided as a 10 mM solution in DMSO (1 ML) and has reported in vitro and cellular activity at micromolar concentrations.
  • Allosteric activator of GPX4 with reported pEC50 4.7.
  • Increases GPX4 enzymatic activity up to ~150% in cell-free assays.
  • Shows cellular activity and protects against cholesterol peroxide toxicity.
  • High reported purity (99.94%) suitable for research use.
  • Supplied as a 10 mM solution in DMSO, ready for in vitro applications.
  • Storage and solubility data provided for experimental planning.
Catalog No. 50-003-07946 Supplier Medchemexpress LLC Supplier No. HY11562710MM 1ML
PKUMDL-LC-101-D04 is a small-molecule allosteric activator of glutathione peroxidase 4 (GPX4) used in research to inhibit ferroptosis and modulate inflammatory responses. The compound is provided as a 10 mM solution in DMSO (1 ML) and has reported in vitro and cellular activity at micromolar concentrations.
  • Allosteric activator of GPX4 with reported pEC50 4.7.
  • Increases GPX4 enzymatic activity up to ~150% in cell-free assays.
  • Shows cellular activity and protects against cholesterol peroxide toxicity.
  • High reported purity (99.94%) suitable for research use.
  • Supplied as a 10 mM solution in DMSO, ready for in vitro applications.
  • Storage and solubility data provided for experimental planning.