Learn More
Medchemexpress LLC PKUMDL-LC-101-D04 1g | 2143896-83-5 | 378.94 | C14H23ClN4O2S2 | 1 G
SDP
Catalog No. 5000226642
Encompass_Preferred
PKUMDL-LC-101-D04 is a small-molecule allosteric activator of glutathione peroxidase 4 (GPX4) supplied as a hydrochloride research chemical. It increases GPX4 enzymatic activity (reported pEC50 = 4.7) and has been used in cell and animal studies to inhibit ferroptosis and reduce inflammation. Molecular formula C14H23ClN4O2S2; molecular weight 378.94 Da.
  • Allosteric activator of glutathione peroxidase 4 (GPX4).
  • Reported activity pEC50 = 4.7 in biochemical assays.
  • Hydrochloride salt form for improved stability and handling.
  • Molecular formula C14H23ClN4O2S2, molecular weight 378.94 Da.
  • Provided as a research reagent; not for human use.
  • Typical pack size: 1 g.
Catalog No. 50-002-26642 Supplier Medchemexpress LLC Supplier No. HY1156271G
PKUMDL-LC-101-D04 is a small-molecule allosteric activator of glutathione peroxidase 4 (GPX4) supplied as a hydrochloride research chemical. It increases GPX4 enzymatic activity (reported pEC50 = 4.7) and has been used in cell and animal studies to inhibit ferroptosis and reduce inflammation. Molecular formula C14H23ClN4O2S2; molecular weight 378.94 Da.
  • Allosteric activator of glutathione peroxidase 4 (GPX4).
  • Reported activity pEC50 = 4.7 in biochemical assays.
  • Hydrochloride salt form for improved stability and handling.
  • Molecular formula C14H23ClN4O2S2, molecular weight 378.94 Da.
  • Provided as a research reagent; not for human use.
  • Typical pack size: 1 g.