Learn More
Apexbio Technology LLC PKUMDL-LC-101-D04 2143896-83-5 25mg
SDP
Catalog No. 5000087923
Encompass_Preferred
PKUMDL-LC-101-D04 (CAS 2143896-83-5) is a small molecule allosteric activator of glutathione peroxidase 4 (GPX4) a key enzyme regulating ferroptosis In biochemical and cell extract assays PKUMDL-LC-101-D04 enhances GPX4 activity to 150% of baseline at concentrations of 20 M and 61 M respectively This compound serves as a valuable tool for investigating the modulation of ferroptosis through GPX4 activation and supports research into oxidative stress and cell death mechanisms
Catalog No. 50-000-87923 Supplier Apexbio Technology LLC Supplier No. BA344225
PKUMDL-LC-101-D04 (CAS 2143896-83-5) is a small molecule allosteric activator of glutathione peroxidase 4 (GPX4) a key enzyme regulating ferroptosis In biochemical and cell extract assays PKUMDL-LC-101-D04 enhances GPX4 activity to 150% of baseline at concentrations of 20 M and 61 M respectively This compound serves as a valuable tool for investigating the modulation of ferroptosis through GPX4 activation and supports research into oxidative stress and cell death mechanisms