Learn More
Medchemexpress LLC PKUMDL-LC-101-D04 250mg | 2143896-83-5 | 378.94 g/mol | C14H23ClN4O2S2 | 250 MG
SDP
Catalog No. 5000228915
Encompass_Preferred
This small-molecule allosteric activator of glutathione peroxidase 4 (GPX4) increases GPX4 enzymatic activity (reported pEC50 = 4.7), inhibits ferroptosis and inflammation in biochemical and cellular assays, and is supplied as a high-purity research reagent for laboratory studies.
  • Activates GPX4 with reported pEC50 = 4.7.
  • Increases GPX4 activity to approximately 150% in cell-free assays.
  • Inhibits ferroptosis and associated inflammatory responses.
  • High purity (≈99.9%) suitable for research applications.
  • Soluble in DMSO at about 200 mg/mL for preparation of stock solutions.
  • White to off-white solid; store sealed and away from moisture.
Catalog No. 50-002-28915 Supplier Medchemexpress LLC Supplier No. HY115627250MG
This small-molecule allosteric activator of glutathione peroxidase 4 (GPX4) increases GPX4 enzymatic activity (reported pEC50 = 4.7), inhibits ferroptosis and inflammation in biochemical and cellular assays, and is supplied as a high-purity research reagent for laboratory studies.
  • Activates GPX4 with reported pEC50 = 4.7.
  • Increases GPX4 activity to approximately 150% in cell-free assays.
  • Inhibits ferroptosis and associated inflammatory responses.
  • High purity (≈99.9%) suitable for research applications.
  • Soluble in DMSO at about 200 mg/mL for preparation of stock solutions.
  • White to off-white solid; store sealed and away from moisture.