Learn More
Medchemexpress LLC PKUMDL-LC-101-D04 50mg | 2143896-83-5 | 378.94 g·mol⁻¹ | C14H23ClN4O2S2 | 50 MG
SDP
Catalog No. 5000228548
Encompass_Preferred
Small-molecule allosteric activator of glutathione peroxidase 4 (GPX4) used as a research tool to study ferroptosis and inflammation. Reported activity includes an approximate pEC50 of 4.7 and an ability to increase GPX4 activity in biochemical assays. Supplied as a solid with good solubility in DMSO for assay preparation.
  • Allosteric activation of glutathione peroxidase 4 with reported pEC50 ≈ 4.7.
  • Shown to increase GPX4 activity to about 150% in biochemical assays.
  • Useful for probing ferroptosis and inflammation pathways in preclinical research.
  • Supplied as a solid with high DMSO solubility for convenient dilution and assay use.
  • Vendor-reported molecular formula and molecular weight provided for analytical reference.
Catalog No. 50-002-28548 Supplier Medchemexpress LLC Supplier No. HY11562750MG
Small-molecule allosteric activator of glutathione peroxidase 4 (GPX4) used as a research tool to study ferroptosis and inflammation. Reported activity includes an approximate pEC50 of 4.7 and an ability to increase GPX4 activity in biochemical assays. Supplied as a solid with good solubility in DMSO for assay preparation.
  • Allosteric activation of glutathione peroxidase 4 with reported pEC50 ≈ 4.7.
  • Shown to increase GPX4 activity to about 150% in biochemical assays.
  • Useful for probing ferroptosis and inflammation pathways in preclinical research.
  • Supplied as a solid with high DMSO solubility for convenient dilution and assay use.
  • Vendor-reported molecular formula and molecular weight provided for analytical reference.