Learn More
Visikol Inc Visikol Plant Biology 100mL
Catalog No. 501616401
Encompass_Preferred
Laboratory Reagent, Histological Reagent, Tissue Clearing Reagent, Visikol for Plant Biology, 100mL, A simple to use rapid tissue clearing formula optimized for botanical microscopy, Clears most tissues within 12 hours, Great for taxanomical morphological and pathological identification, Comes in a white HDPE bottle with black cap.
Catalog No. 50-161-6401 Supplier Visikol Inc Supplier No. OV100
Laboratory Reagent, Histological Reagent, Tissue Clearing Reagent, Visikol for Plant Biology, 100mL, A simple to use rapid tissue clearing formula optimized for botanical microscopy, Clears most tissues within 12 hours, Great for taxanomical morphological and pathological identification, Comes in a white HDPE bottle with black cap.