Learn More
Eurofins Discoverx ProLabel Cloning Vector Bundle, 2 vials, 10 µg. (93-1109)
Catalog No. 502002128
Encompass
The ProLabel Cloning Vector Bundle is a bundle of two mammalian cloning vectors (C and N terminal tags) for use in expressing a protein of interest fused to the ProLabel (ProLabel) tag. The ProLabel tag is a version of ED used for EFC assays (in combination with EA) where ED and EA are contained in separate cellular compartments. Provided as two vials, 1 vial each vector, of 10 μg vector in 20 μL liquid buffer. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/reagent/vector/cloning-vector-bundle-93-1109.
Catalog No. 50-200-2128 Supplier Eurofins Discoverx Supplier No. 931109
The ProLabel Cloning Vector Bundle is a bundle of two mammalian cloning vectors (C and N terminal tags) for use in expressing a protein of interest fused to the ProLabel (ProLabel) tag. The ProLabel tag is a version of ED used for EFC assays (in combination with EA) where ED and EA are contained in separate cellular compartments. Provided as two vials, 1 vial each vector, of 10 μg vector in 20 μL liquid buffer. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/reagent/vector/cloning-vector-bundle-93-1109.