Learn More
Chemglass Life Sciences 2000mL Single Neck Evaporating Flask, 24/40 Outer Joint, Plastic Coated Recovery Flask
SDP
Catalog No. CG1512P35
Encompass_Preferred
2000mL Single Neck Evaporating Flask, 24/40 Outer Joint, Plastic Coated Recovery Flask. Similar to CG-1512 recovery flasks, but plastic coated to prevent injury from broken glass and spillage of hazardous materials.
Catalog No. CG1512P35 Supplier Chemglass Life Sciences Supplier No. CG1512P35
2000mL Single Neck Evaporating Flask, 24/40 Outer Joint, Plastic Coated Recovery Flask. Similar to CG-1512 recovery flasks, but plastic coated to prevent injury from broken glass and spillage of hazardous materials.