Learn More
Med Vet International Jorvet Ketch-All Pole, 4-6ft, J0915B, Each
Catalog No. 5000495844
Encompass_Preferred
This is the original "Rabies Stick" for restraint of vicious or hard to control animals. The metal pole has a locking mechanism to hold the cable tight plus a quick release button. Rubber cushion grips for two-handed no slip control. The end of the shaft has a covered portion to protect against chewing. Ketch-All Pole is 4-6ft. Suitable for cats and dogs.
Catalog No. 50-004-95844 Supplier Med Vet International Supplier No. JOR915B
This is the original "Rabies Stick" for restraint of vicious or hard to control animals. The metal pole has a locking mechanism to hold the cable tight plus a quick release button. Rubber cushion grips for two-handed no slip control. The end of the shaft has a covered portion to protect against chewing. Ketch-All Pole is 4-6ft. Suitable for cats and dogs.