Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ Quick Gel Extraction and PCR Purification Combo Kit

Catalog No. k220001
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K220001 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K220001 Supplier Invitrogen™ Supplier No. K220001
Only null left
Add to Cart
Edge
Add to Cart

Includes

The PureLink Quick Gel Extraction & PCR Purification Combo Kit includes sufficient PureLink columns, collection tubes, elution tubes, and reagents for 50 preps.

Offers ability to perform both a gel extraction or PCR purification with single kit.

PureLink™ Quick Gel Extraction and PCR Purification Combo Kit offers the ability to perform both a gel extraction or a PCR purification a single kit.

The PureLink™ Quick Gel Extraction and PCR Purification Combo Kit is designed to purify DNA fragments from agarose gels. The simple procedure uses a silica-based spin cartridge to purify DNA fragments from 40 bp to 10 kb in < 30 minutes. It also provide all components to perform a rapid and efficient removal of short primers, dNTPs, enzymes, short-failed PCR products, and salts from PCR fragments >100 bp in less than 10 minutes. Isolated DNA using the PureLink™ kit is free of proteins, dye, and agarose, and is ready-to-use for a variety of applications, including automated fluorescent DNA sequencing, manual DNA sequencing, PCR, in vitro transcription, restriction mapping, cloning, and labeling. The kit contains all the buffers from the PureLink™ Quick Gel extraction kit and the PCR binding buffer from PureLink™ PCR Purification kit with Invitrogen branded lidded column.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.
Isolation Technology Silica Spin Column
Scale Mini
Sample Type Linear DNA fragments from: Agarose Gels, PCR Purification, Restriction Enzyme Reactions, PCR Products
Elution Volume 50 μL
Final Product Type DNA
For Use With (Application) PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput Compatibility Not High-throughput Compatible (Manual)
Label or Dye Ethidium Bromide, SYBR Safe
Quantity 50 Preps
Shipping Condition Room Temperature
Test Time Up to 30 min.
Yield 15 μg (Binding capacity)
Starting Material Amount Agarose Gel: ≤400 mg, PCR Product: ≤100 μL, or ≤40 μg dsDNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.